Learn More
Integra Biosciences Corp Tube Rack For 2 ml HPLC Vials, 6x8
SDP
Catalog No. NC3242507
Encompass
Compatible with 4, 6 or 8 channel VOYAGER tip spacing pipettes and D-ONE pipetting moduleStores up to 48 x 2.0 ml HPLC vialsLabware Format 6x8SLAS/ANSI footprint
Catalog No. NC3242507 Supplier Integra Biosciences Corp Supplier No. 4545
Compatible with 4, 6 or 8 channel VOYAGER tip spacing pipettes and D-ONE pipetting moduleStores up to 48 x 2.0 ml HPLC vialsLabware Format 6x8SLAS/ANSI footprint