Learn More
V&P Scientific Inc TUBE RACK for 15 x 92mm Diameter Sample Tubes with Labels, 4 x 6
Catalog No. NC1849302
Encompass
TUBE RACK, 2 3D-printed PLA Plates, Holds 15 x 92mm Plastic, Flat Bottom Sample Tubes with Labels, 4 x 6 for 24 Total, 6.4mm Thick Top and Bottom Plate, Connected with Stainless Steel Stand-offs, for 80mm Overall Height, SLAS Footprint
Catalog No. NC1849302 Supplier V&P Scientific Inc Supplier No. VP 4182415PLA
TUBE RACK, 2 3D-printed PLA Plates, Holds 15 x 92mm Plastic, Flat Bottom Sample Tubes with Labels, 4 x 6 for 24 Total, 6.4mm Thick Top and Bottom Plate, Connected with Stainless Steel Stand-offs, for 80mm Overall Height, SLAS Footprint