Learn More
Alkali Scientific Two-teired Tube Rack for 16 X 5mL MacroTube, Package of 1 Rack
SDP
Catalog No. 502389196
Encompass_Preferred
A great workstation Tube Rack for 16 x 5mL MacroTube, designed specifically for 5mL MacroTube (also accepts Eppendorf). Tubes are held securely in custom shaped wells that accepts 5mL snap cap and 5mL screw cap tubes. Workstation is also perfect for 15mL conical bottom tubes. Two tiers make it easier to pipette into the second row and easier to see which tubes are filled or empty.
Catalog No. 50-238-9196 Supplier Alkali Scientific Supplier No. C2590
A great workstation Tube Rack for 16 x 5mL MacroTube, designed specifically for 5mL MacroTube (also accepts Eppendorf). Tubes are held securely in custom shaped wells that accepts 5mL snap cap and 5mL screw cap tubes. Workstation is also perfect for 15mL conical bottom tubes. Two tiers make it easier to pipette into the second row and easier to see which tubes are filled or empty.