Learn More
Research Products International Corp Tube Rack for Magnetic Platform
SDP
Catalog No. 501977654
Encompass
MAGic Clamp Rack is for use with Incu-Shaker Mini and Incu-Shaker 10L model Shaking Incubators.
Magnetic tube rack secures test tubes to a shaker platform for mixing or aerating operations.
Catalog No. 50-197-7654 Supplier Research Products International Corp Supplier No. 248577
MAGic Clamp Rack is for use with Incu-Shaker Mini and Incu-Shaker 10L model Shaking Incubators.
Magnetic tube rack secures test tubes to a shaker platform for mixing or aerating operations.