Learn More
Electron Microscopy Sciences Multi-tier tube rack, blue
SDP
Catalog No. 501928304
Encompass_Preferred
This multi-tier tube rack has four rows of holes arranged at four different levels. The highest row can accommodate five 30 or 50 ml centrifuge tubes in either the conical or freestanding configuration. The second row can hold eight 15 ml centrifuge tubes. The third row has ten holes for 12 x 75 mm tubes. The front row has 12 holes for microcentrifuge tubes.
Specially designed for applications where space is limited, but multiple size racks are required. Made of a heavy-duty, high impact polymer; this rack is truly versatile.
Color: blue
Catalog No. 50-192-8304 Supplier Electron Microscopy Sciences Supplier No. 7236601
This multi-tier tube rack has four rows of holes arranged at four different levels. The highest row can accommodate five 30 or 50 ml centrifuge tubes in either the conical or freestanding configuration. The second row can hold eight 15 ml centrifuge tubes. The third row has ten holes for 12 x 75 mm tubes. The front row has 12 holes for microcentrifuge tubes.
Specially designed for applications where space is limited, but multiple size racks are required. Made of a heavy-duty, high impact polymer; this rack is truly versatile.
Color: blue