Learn More
Chemglass Life Sciences 100mL Tube, Reaction, 24/40 Outer Joint, Valve, Airfree, Schlenk
SDP
Catalog No. 501530970
Encompass_Preferred
100mL Tube, Reaction, 24/40 Outer Joint, Valve, Airfree, Schlenk. Cylindrical shaped tube useful for storage or simple reactions. Tube has a standard taper outer joint. Available with a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. 50-153-0970 Supplier Chemglass Life Sciences Supplier No. AF053724
100mL Tube, Reaction, 24/40 Outer Joint, Valve, Airfree, Schlenk. Cylindrical shaped tube useful for storage or simple reactions. Tube has a standard taper outer joint. Available with a 0-4mm Chem-Cap Airfree inlet valve.