Learn More
Chemglass Life Sciences 50mL Tube, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk
SDP
Catalog No. 50873471
Encompass_Preferred
50mL Tube, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk. Cylindrical shaped tube useful for storage or simple reactions. Tube has a standard taper outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.
Catalog No. 50-873-471 Supplier Chemglass Life Sciences Supplier No. AF053721
50mL Tube, Reaction, 14/20 Outer Joint, Valve, Airfree, Schlenk. Cylindrical shaped tube useful for storage or simple reactions. Tube has a standard taper outer joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve.