Learn More
Chemglass Life Sciences 1mL Tube, Recrystallization Tube, Minum-Ware
SDP
Catalog No. 501532700
Encompass_Preferred
1mL Tube, Recrystallization Tube, Minum-Ware. Used for microscale recrystallizing of small quantities of reaction product. Consists of a glass outer tube with a non-uniform grind to purposely allow leakage, and a PTFE inner plug. The PTFE inner plug fits both the 1.0ml and 2.0ml capacity outer tubes. Capacity listed is the volume of the tube below the ground seat.
Catalog No. 50-153-2700 Supplier Chemglass Life Sciences Supplier No. MW7403
1mL Tube, Recrystallization Tube, Minum-Ware. Used for microscale recrystallizing of small quantities of reaction product. Consists of a glass outer tube with a non-uniform grind to purposely allow leakage, and a PTFE inner plug. The PTFE inner plug fits both the 1.0ml and 2.0ml capacity outer tubes. Capacity listed is the volume of the tube below the ground seat.