Learn More
Chemglass Life Sciences PTFE Plunger Only
SDP
Catalog No. 501532702
Encompass_Preferred
PTFE Plunger Only. The PTFE inner plug fits both the 1.0ml and 2.0ml capacity outer tubes.
Catalog No. 50-153-2702 Supplier Chemglass Life Sciences Supplier No. MW7405
PTFE Plunger Only. The PTFE inner plug fits both the 1.0ml and 2.0ml capacity outer tubes.