Learn More
V&P Scientific Inc MAGNETIC RACK, For use with the VP 772F2M-1 Base, 14 Flip Top Microcentrifuge Tubes
Catalog No. NC0542877
Encompass
MAGNETIC BEAD SEPARATION RACK, For use with the VP 772F2M-1 Base, 14 Flip Top Microcentrifuge Tubes,1.5mL or 5.0mL Volume, 1 Sided Bead Pellet Location, Variable Positions, 48MGO, NdFeB Magnet, 18mm Center Spacing, 13mm Hole Diameter, Polycarbonate
Catalog No. NC0542877 Supplier V&P Scientific Inc Supplier No. VP 772F2R1
MAGNETIC BEAD SEPARATION RACK, For use with the VP 772F2M-1 Base, 14 Flip Top Microcentrifuge Tubes,1.5mL or 5.0mL Volume, 1 Sided Bead Pellet Location, Variable Positions, 48MGO, NdFeB Magnet, 18mm Center Spacing, 13mm Hole Diameter, Polycarbonate