Learn More
Thomas Scientific Test Tube, 13 x 100mm (8mL), PS, 1000 case
SDP
Catalog No. NC1798670
Encompass
Globe Scientific offers an extensive line of mulit-use test tubes. They are molded from premium grade polystyrene for clarity or of polypropylene for impact strength and stability to common acids, alkalies, and solvents. Polystyrene tubes can withstand 1500 RPM during centrifugation. Polypropylene tubes can withstand 3000 RPM and are autoclavable and freezeable. Globe's broad range of sizes are sure to fit most laboratory applications.
Catalog No. NC1798670 Supplier Thomas Scientific Supplier No. 1198X99
Globe Scientific offers an extensive line of mulit-use test tubes. They are molded from premium grade polystyrene for clarity or of polypropylene for impact strength and stability to common acids, alkalies, and solvents. Polystyrene tubes can withstand 1500 RPM during centrifugation. Polypropylene tubes can withstand 3000 RPM and are autoclavable and freezeable. Globe's broad range of sizes are sure to fit most laboratory applications.