Learn More
Electron Microscopy Sciences Tube Rocker, 1/EA, 230V
SDP
Catalog No. 502090889
Encompass_Preferred
The TubeRocker is designed to provide the ideal speed and tilt angle for the agitation of sample tubes. The grooved design of the included rubber mat properly secures all tube sizes ranging from 1.5 to 50 ml, without the need for added straps or bands. Small 5 x 13" size makes it easy to fit on any lab bench. It can be conveniently placed inside refrigerators, cold rooms or laboratory incubators. Simple-to-use. Needs no adapters or mounting hardware. Just press the power switch and let it rock!
Catalog No. 50-209-0889 Supplier Electron Microscopy Sciences Supplier No. 6570012
The TubeRocker is designed to provide the ideal speed and tilt angle for the agitation of sample tubes. The grooved design of the included rubber mat properly secures all tube sizes ranging from 1.5 to 50 ml, without the need for added straps or bands. Small 5 x 13" size makes it easy to fit on any lab bench. It can be conveniently placed inside refrigerators, cold rooms or laboratory incubators. Simple-to-use. Needs no adapters or mounting hardware. Just press the power switch and let it rock!