Learn More
Thermo Scientific™ Tube Rocker Stacking Bars
Catalog No. 88882160 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
88-882-160 Stainless Steel
1 options
Catalog No. 88-882-160 Supplier Thermo Scientific™ Supplier No. 88882160
  • $108.10
    $83.65 / Each of 1
    Save $24.45
    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Accessory for use with Digital Tube Rocker, 120V and Digital Tube Rocker, 230V
Double your capacity in a compact footprint while using the Thermo Scientific™ Digital Tube Rocker with the Tube Rocker Stacking Bars.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Dimensions (L x W) 0.8 x 6 cm
Description Tube Rocker Stacking Bars
For Use With (Application) Platform Tray (88882158), Tube Rocker Stacking Bars (88882160), Variety of tubes
For Use With (Equipment) Digital Tube Rocker, 120V (88882017), Digital Tube Rocker 230V (88882018)
No. of Positions 4 stacking bars for 2nd tube rocker tray installation