Learn More
Research Products International Corp Tube Rocker with Grooved Mat, Accepts 1.5ml Micro-tubes up to 50ml Centrifuge Tubes
SDP
Catalog No. 50998800
Encompass
Tube Rocker is designed to provide ideal speed and tilt angle for agitation of tubes in a variety of laboratory applications. Tube Rocker accepts 1.5ml micro-tubes up to 50ml centrifuge tubes. Grooved design of rubber mat properly secures all tubes. Compact size (5" x 13") fits conveniently on lab bench, incubator, refrigerator or cold room.
Catalog No. 50-998-800 Supplier Research Products International Corp Supplier No. 246261
Tube Rocker is designed to provide ideal speed and tilt angle for agitation of tubes in a variety of laboratory applications. Tube Rocker accepts 1.5ml micro-tubes up to 50ml centrifuge tubes. Grooved design of rubber mat properly secures all tubes. Compact size (5" x 13") fits conveniently on lab bench, incubator, refrigerator or cold room.