Learn More
Cole-Parmer Boekel Scientific 260650 Tube Rotator, 100-240V
Supplier:  Cole-Parmer 5120200
Encompass
This tube rotator features a digital display and microprocessor speed control from 4 to 80 rpm — ensuring repeatable mixing actions from gentle to vigorous. Carousel head is adjustable from 90° to 180° to offer different mixing angles. Choice of five rotor attachments provides great versatility for most mixing needs; appropriate clips are included. Digital timer can count down or count up.
Catalog No. NC0766137