Learn More
Alkali Scientific Tube Revolver with Variable Speed
SDP
Catalog No. 502388804
Encompass_Preferred
The Tube Revolver can be used in a variety of laboratory fields such as Molecular Biology, Biochemistry, Clinical, and Histochemistry. The rotisseries are interchangeable so that the unit works with a variety of tube sizes. The four rotisserie paddles can be mixed for a combination of different tubes. Each paddle direction can be independently adjusted form 0-90 degrees. 2 Year Warranty Accessories in standard package: Rotisserie Paddles for 10mL/15mL Tubes (Qty 2) Rotisserie Paddles for 5mL/7mL Tubes (Qty 2) Rotisserie Paddles for 1.5mL/2mL Tubes (Qty 2) Rotisserie Paddles for 0.5mL/0.8mL Tubes (Qty 2) Rotisserie Paddles for 50mL Tubes (Qty 1)
Catalog No. 50-238-8804 Supplier Alkali Scientific Supplier No. RS3552A
The Tube Revolver can be used in a variety of laboratory fields such as Molecular Biology, Biochemistry, Clinical, and Histochemistry. The rotisseries are interchangeable so that the unit works with a variety of tube sizes. The four rotisserie paddles can be mixed for a combination of different tubes. Each paddle direction can be independently adjusted form 0-90 degrees. 2 Year Warranty Accessories in standard package: Rotisserie Paddles for 10mL/15mL Tubes (Qty 2) Rotisserie Paddles for 5mL/7mL Tubes (Qty 2) Rotisserie Paddles for 1.5mL/2mL Tubes (Qty 2) Rotisserie Paddles for 0.5mL/0.8mL Tubes (Qty 2) Rotisserie Paddles for 50mL Tubes (Qty 1)