Learn More
Chemglass Life Sciences Tube for Sample Decomposition, Ø22mm (For COD analysis only)
SDP
Catalog No. 5000493101
Encompass_Preferred
Tube for Sample Decomposition, Ø22mm (For COD analysis only). For use with CG-1991-V COD ECODryBlock Systems.
Catalog No. 50-004-93101 Supplier Chemglass Life Sciences Supplier No. CG1991V214
Tube for Sample Decomposition, Ø22mm (For COD analysis only). For use with CG-1991-V COD ECODryBlock Systems.