Learn More
Chemglass Life Sciences Tube, Sample, NMR, 5mm, With Caps, 7" Long, 500 MHz
SDP
Catalog No. CS5500QTZ7
Encompass_Preferred
Tube, Sample, NMR, 5mm, With Caps, 7" Long, 500 MHz. NORELL Thin-Wall Quartz Glass NMR Sample Tubes are clear fused quartz, 99.8% SiO2. Containing < 0.1ppm Boron. Use these tubes for applications requiring temperatures above 150¿C & UV irradiation studies (Greater than 80% T above 255nm).
Catalog No. CS5500QTZ7 Supplier Chemglass Life Sciences Supplier No. CS5500QTZ7
Tube, Sample, NMR, 5mm, With Caps, 7" Long, 500 MHz. NORELL Thin-Wall Quartz Glass NMR Sample Tubes are clear fused quartz, 99.8% SiO2. Containing < 0.1ppm Boron. Use these tubes for applications requiring temperatures above 150¿C & UV irradiation studies (Greater than 80% T above 255nm).