Learn More
Med Vet International AMSure Suction Tubing, 0.188 Inch Inner Diameter, 6 Foot Length, Sterile, AS821, Each
Catalog No. 503044441
Encompass_Preferred
0.188-inch inner diameter with a 2-yard length The tubing has a molded female connector on both ends; there is one male-male connector inserted onto one end in case healthcare worker wants to inter-connect more than one piece. Flexible, low coil memory allows for easy draping. Collapse-resistant striated tubing improves clinical outcomes. Green molded female connectors deliver secure connections to reduce procedure interruptions.
Catalog No. 50-304-4441 Supplier Med Vet International Supplier No. 483597EA
0.188-inch inner diameter with a 2-yard length The tubing has a molded female connector on both ends; there is one male-male connector inserted onto one end in case healthcare worker wants to inter-connect more than one piece. Flexible, low coil memory allows for easy draping. Collapse-resistant striated tubing improves clinical outcomes. Green molded female connectors deliver secure connections to reduce procedure interruptions.