Learn More
Thomas Scientific Centrifuge Tube, 50mL, PP, Free Standing, Flat Screw Cap, 25/sterile bag
SDP
Catalog No. 501458007
Encompass_Preferred
Conical 15mL and 50mL centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86°C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Sterile, RNase, DNase, pyrogen and cytotoxin free Available racked or bagged Compatible with standard rotors Frosted marking surface *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions and adapters.
Catalog No. 50-145-8007 Supplier Thomas Scientific Supplier No. 1158Q87
Conical 15mL and 50mL centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86°C. The maximum centrifugal force is 12,000xg for the 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Sterile, RNase, DNase, pyrogen and cytotoxin free Available racked or bagged Compatible with standard rotors Frosted marking surface *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions and adapters.