Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .020in ID silicone with male and female luers. Package of 5, nonsterile.
SDP
Catalog No. 5081187
Encompass
The Instech P720/TS‑FL020SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, built with .020in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets provide secure, low‑dead‑volume connections for precise low‑flow infusion and perfusion applications.
Features
  • .020in ID silicone tubing for low‑flow P720 operation
  • Female luer inlet and male luer outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for controlled infusion and general research fluid handling
Catalog No. 50-811-87 Supplier Instech Supplier No. P720/TSFL020SLL
The Instech P720/TS‑FL020SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, built with .020in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets provide secure, low‑dead‑volume connections for precise low‑flow infusion and perfusion applications.
Features
  • .020in ID silicone tubing for low‑flow P720 operation
  • Female luer inlet and male luer outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for controlled infusion and general research fluid handling