Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .031in ID silicone with male and female luers. Package of 5, nonsterile.
SDP
Catalog No. 5081188
Encompass
The Instech P720/TS‑FL031SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, made with .031in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets provide secure, low‑dead‑volume connections suitable for mid‑range low‑flow infusion and perfusion applications.
Features
  • .031in ID silicone tubing for mid‑range P720 flow rates
  • Female luer inlet and male luer outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for precise infusion, perfusion, and general research workflows
Catalog No. 50-811-88 Supplier Instech Supplier No. P720/TSFL031SLL
Add to Cart
Add to Cart
The Instech P720/TS‑FL031SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, made with .031in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets provide secure, low‑dead‑volume connections suitable for mid‑range low‑flow infusion and perfusion applications.
Features
  • .031in ID silicone tubing for mid‑range P720 flow rates
  • Female luer inlet and male luer outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for precise infusion, perfusion, and general research workflows