Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .062in ID silicone with male and female luers. Package of 5, nonsterile.
SDP
Catalog No. 5081189
Encompass
The Instech P720/TS‑FL062SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, built with .062in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets support higher low‑flow and mid‑flow applications while providing secure, low‑dead‑volume luer connections for research infusion workflows.
Features
  • .062in ID silicone tubing for higher low‑flow ranges
  • Female luer inlet and male luer outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 peristaltic pump models
  • Suitable for infusion, perfusion, and general research applications
Catalog No. 50-811-89 Supplier Instech Supplier No. P720/TSFL062SLL
The Instech P720/TS‑FL062SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, built with .062in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets support higher low‑flow and mid‑flow applications while providing secure, low‑dead‑volume luer connections for research infusion workflows.
Features
  • .062in ID silicone tubing for higher low‑flow ranges
  • Female luer inlet and male luer outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 peristaltic pump models
  • Suitable for infusion, perfusion, and general research applications