Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .093in ID silicone with male and female luers. Package of 5, nonsterile.
SDP
Catalog No. 5081190
Encompass
The Instech P720/TS‑FL093SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, built with .093in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets provide secure, low‑dead‑volume connections suitable for high‑flow applications within the P720 operating range.
Features
  • .093in ID silicone tubing for high‑flow P720 performance
  • Female luer inlet and male luer outlet connectors
  • Non‑sterile tube sets supplied in packs of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for infusion, perfusion, and general lab fluid handling
Catalog No. 50-811-90 Supplier Instech Supplier No. P720/TSFL093SLL
The Instech P720/TS‑FL093SLL is a luer‑to‑luer tube set for P720 peristaltic pumps, built with .093in ID silicone tubing and fitted with a female luer on one end and a male luer on the other. Supplied non‑sterile in packs of five, these tube sets provide secure, low‑dead‑volume connections suitable for high‑flow applications within the P720 operating range.
Features
  • .093in ID silicone tubing for high‑flow P720 performance
  • Female luer inlet and male luer outlet connectors
  • Non‑sterile tube sets supplied in packs of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for infusion, perfusion, and general lab fluid handling