Learn More
Instech Tube sets for Instech P720 peristaltic pumps, dual channel, .031in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing). Package of 5, nonsterile.
SDP
Catalog No. 5081183
Encompass
The Instech P720/TS‑DBS031SDBS is a dual‑barb tube set for P720 peristaltic pumps, built with .031in ID silicone tubing in a two‑channel configuration. Designed for applications requiring simultaneous or parallel fluid paths, these non‑sterile tube sets feature .062in ID dual barb connectors on each end and are supplied in packs of five for reliable multi‑channel low‑flow delivery.
Features
  • Two‑channel silicone tube set for P720 pumps
  • .031in ID tubing for mid‑range low‑flow rates
  • Dual .062in ID barb connectors on each end
  • Non‑sterile sets packaged in packs of five
  • Supports parallel or dual‑path infusion and perfusion
Catalog No. 50-811-83 Supplier Instech Supplier No. P720/TSDBS031SDBS
The Instech P720/TS‑DBS031SDBS is a dual‑barb tube set for P720 peristaltic pumps, built with .031in ID silicone tubing in a two‑channel configuration. Designed for applications requiring simultaneous or parallel fluid paths, these non‑sterile tube sets feature .062in ID dual barb connectors on each end and are supplied in packs of five for reliable multi‑channel low‑flow delivery.
Features
  • Two‑channel silicone tube set for P720 pumps
  • .031in ID tubing for mid‑range low‑flow rates
  • Dual .062in ID barb connectors on each end
  • Non‑sterile sets packaged in packs of five
  • Supports parallel or dual‑path infusion and perfusion