Learn More
Instech Tube sets for Instech P720 peristaltic pumps, dual channel, .031in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing), unbalanced for maintaining levels in chambers. Package of 5, nonsterile.
SDP
Catalog No. 5081184
Encompass
The Instech P720/TS‑DBS031SDBSU is an unbalanced two‑channel tube set for P720 peristaltic pumps, built with .031in ID silicone tubing and dual .062in barb connectors on each end. Designed for differential or asymmetric flow conditions, these non‑sterile tube sets are supplied in packs of five and support smooth, reliable dual‑path fluid delivery in research applications.
Features
  • Unbalanced two‑channel configuration for differential flow
  • .031in ID silicone tubing for mid‑range flow rates
  • Dual .062in barb inlet and outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 two‑channel pump configurations
Catalog No. 50-811-84 Supplier Instech Supplier No. P720/TSDBS031SDBSU
The Instech P720/TS‑DBS031SDBSU is an unbalanced two‑channel tube set for P720 peristaltic pumps, built with .031in ID silicone tubing and dual .062in barb connectors on each end. Designed for differential or asymmetric flow conditions, these non‑sterile tube sets are supplied in packs of five and support smooth, reliable dual‑path fluid delivery in research applications.
Features
  • Unbalanced two‑channel configuration for differential flow
  • .031in ID silicone tubing for mid‑range flow rates
  • Dual .062in barb inlet and outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 two‑channel pump configurations