Learn More
Instech Tube sets for Instech P720 peristaltic pumps, dual channel, .062in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing). Package of 5, nonsterile. Warning: will place additional stress on motor and shorten life of the pump.
SDP
Catalog No. 5081185
Encompass
The Instech P720/TS‑DBS062SDBS is a dual‑channel tube set for P720 peristaltic pumps, built with .062in ID silicone tubing and dual .062in barb connectors on each end for parallel high‑flow delivery. Supplied non‑sterile in packs of five, these tube sets support smooth, balanced two‑channel fluid transfer for infusion, perfusion, and other research applications.
Features
  • Two‑channel silicone tube set for P720 pumps
  • .062in ID tubing for higher low‑flow and mid‑flow ranges
  • Dual .062in barb inlet and outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 two‑channel configurations
Catalog No. 50-811-85 Supplier Instech Supplier No. P720/TSDBS062SDBS
The Instech P720/TS‑DBS062SDBS is a dual‑channel tube set for P720 peristaltic pumps, built with .062in ID silicone tubing and dual .062in barb connectors on each end for parallel high‑flow delivery. Supplied non‑sterile in packs of five, these tube sets support smooth, balanced two‑channel fluid transfer for infusion, perfusion, and other research applications.
Features
  • Two‑channel silicone tube set for P720 pumps
  • .062in ID tubing for higher low‑flow and mid‑flow ranges
  • Dual .062in barb inlet and outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with all P720 two‑channel configurations