Learn More
Instech Tube sets for Instech P720 peristaltic pumps, dual channel, .062in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing), unbalanced for maintaining levels in chambers. Package of 5, nonsterile.
SDP
Catalog No. 5081186
Encompass
The Instech P720/TS‑DBS062SDBSU is an unbalanced two‑channel tube set for P720 peristaltic pumps, using .062in ID silicone tubing with dual .062in barb connectors on each end. Designed for applications requiring asymmetric channel resistance or differential flow, these non‑sterile tube sets are supplied in packs of five and support smooth, reliable dual‑path fluid delivery in research workflows.
Features
  • Unbalanced dual‑channel silicone tube set
  • .062in ID tubing for higher low‑flow and mid‑flow ranges
  • Dual .062in barb inlet and outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with P720 two‑channel pump configurations
Catalog No. 50-811-86 Supplier Instech Supplier No. P720/TSDBS062SDBSU
The Instech P720/TS‑DBS062SDBSU is an unbalanced two‑channel tube set for P720 peristaltic pumps, using .062in ID silicone tubing with dual .062in barb connectors on each end. Designed for applications requiring asymmetric channel resistance or differential flow, these non‑sterile tube sets are supplied in packs of five and support smooth, reliable dual‑path fluid delivery in research workflows.
Features
  • Unbalanced dual‑channel silicone tube set
  • .062in ID tubing for higher low‑flow and mid‑flow ranges
  • Dual .062in barb inlet and outlet connectors
  • Packaged non‑sterile in sets of five
  • Compatible with P720 two‑channel pump configurations