Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .020in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing). Package of 5, nonsterile.
SDP
Catalog No. 5081179
Encompass
The Instech P720/TS‑BS020SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, built with .020in ID silicone tubing and .062in ID barb connectors for secure integration with standard laboratory tubing. Supplied in non‑sterile packs of five, these tube sets provide reliable, smooth fluid delivery for low‑flow infusion and perfusion applications.
Features
  • .020in ID silicone tubing for low‑flow P720 operation
  • .062in ID barb inlet and outlet connectors
  • Non‑sterile tube sets packaged in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for infusion, perfusion, and general research fluid handling
Catalog No. 50-811-79 Supplier Instech Supplier No. P720/TSBS020SBS
The Instech P720/TS‑BS020SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, built with .020in ID silicone tubing and .062in ID barb connectors for secure integration with standard laboratory tubing. Supplied in non‑sterile packs of five, these tube sets provide reliable, smooth fluid delivery for low‑flow infusion and perfusion applications.
Features
  • .020in ID silicone tubing for low‑flow P720 operation
  • .062in ID barb inlet and outlet connectors
  • Non‑sterile tube sets packaged in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for infusion, perfusion, and general research fluid handling