Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .031in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing). Package of 5, nonsterile.
SDP
Catalog No. 5081180
Encompass
The Instech P720/TS‑BS031SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, built with .031in ID silicone tubing and .062in ID barb connectors to integrate easily with standard research tubing. Supplied non‑sterile in packs of five, these tube sets provide smooth, accurate fluid delivery for mid‑range low‑flow infusion and perfusion applications.
Features
  • .031in ID silicone tubing for mid‑range P720 flow rates
  • .062in ID barb inlet and outlet connectors
  • Packaged in sets of five, non‑sterile
  • Compatible with all P720 peristaltic pump models
  • Ideal for controlled infusion and perfusion tasks
Catalog No. 50-811-80 Supplier Instech Supplier No. P720/TSBS031SBS
The Instech P720/TS‑BS031SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, built with .031in ID silicone tubing and .062in ID barb connectors to integrate easily with standard research tubing. Supplied non‑sterile in packs of five, these tube sets provide smooth, accurate fluid delivery for mid‑range low‑flow infusion and perfusion applications.
Features
  • .031in ID silicone tubing for mid‑range P720 flow rates
  • .062in ID barb inlet and outlet connectors
  • Packaged in sets of five, non‑sterile
  • Compatible with all P720 peristaltic pump models
  • Ideal for controlled infusion and perfusion tasks