Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .062in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing). Package of 5, nonsterile.
SDP
Catalog No. 5081181
Encompass
The Instech P720/TS‑BS062SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, constructed with .062in ID silicone tubing and .062in ID barb connectors to integrate easily with standard research tubing. Supplied non‑sterile in packs of five, these tube sets support smooth, accurate fluid delivery for higher low‑flow applications within the P720 operating range.
Features
  • .062in ID silicone tubing for higher low‑flow rates
  • .062in ID barb inlet and outlet connectors
  • Non‑sterile tube sets packaged in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for infusion, perfusion, and general research fluid handling
Catalog No. 50-811-81 Supplier Instech Supplier No. P720/TSBS062SBS
The Instech P720/TS‑BS062SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, constructed with .062in ID silicone tubing and .062in ID barb connectors to integrate easily with standard research tubing. Supplied non‑sterile in packs of five, these tube sets support smooth, accurate fluid delivery for higher low‑flow applications within the P720 operating range.
Features
  • .062in ID silicone tubing for higher low‑flow rates
  • .062in ID barb inlet and outlet connectors
  • Non‑sterile tube sets packaged in sets of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for infusion, perfusion, and general research fluid handling