Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .093in ID silicone with .062in ID barb inlet and outlet (mates with 062 tubing). Package of 5, nonsterile.
SDP
Catalog No. 5081182
Encompass
The Instech P720/TS‑BS062SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, made with .062in ID silicone tubing and .062in ID barb connectors for secure integration with standard laboratory tubing. Supplied non‑sterile in packs of five, these tube sets provide smooth, reliable fluid delivery for higher low‑flow infusion and perfusion applications.
Features
  • .062in ID silicone tubing for higher low‑flow performance
  • .062in ID barb inlet and outlet connectors
  • Non‑sterile tube sets supplied in packs of five
  • Compatible with all P720 peristaltic pump models
  • Suitable for infusion, perfusion, and general research fluid transfer
Catalog No. 50-811-82 Supplier Instech Supplier No. P720/TSBS093SBS
The Instech P720/TS‑BS062SBS is a barb‑to‑barb tube set for P720 peristaltic pumps, made with .062in ID silicone tubing and .062in ID barb connectors for secure integration with standard laboratory tubing. Supplied non‑sterile in packs of five, these tube sets provide smooth, reliable fluid delivery for higher low‑flow infusion and perfusion applications.
Features
  • .062in ID silicone tubing for higher low‑flow performance
  • .062in ID barb inlet and outlet connectors
  • Non‑sterile tube sets supplied in packs of five
  • Compatible with all P720 peristaltic pump models
  • Suitable for infusion, perfusion, and general research fluid transfer