Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .020in ID silicone, straight through configuration (P720/NCA fittings required), 1ft (30cm) long. Package of 5, nonsterile.
SDP
Catalog No. 5081175
Encompass
The Instech P720/TS‑020S is a 1ft (30cm)straight‑through tube set for P720 peristaltic pumps, built with .020in ID silicone tubing for precise low‑flow delivery. Supplied in packs of five, these non‑sterile tube sets provide connector‑free inlet and outlet routing when used with P720/NCA fittings, supporting accurate and smooth operation in research infusion and perfusion setups.
Features
  • 1ft (30cm) length for flexible pump integration
  • .020in ID silicone tubing for low‑flow pumping
  • Straight‑through design for use with P720/NCA fittings
  • Non‑sterile tube sets supplied in packs of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for precise infusion and perfusion applications
Catalog No. 50-811-75 Supplier Instech Supplier No. P720/TS020S
The Instech P720/TS‑020S is a 1ft (30cm)straight‑through tube set for P720 peristaltic pumps, built with .020in ID silicone tubing for precise low‑flow delivery. Supplied in packs of five, these non‑sterile tube sets provide connector‑free inlet and outlet routing when used with P720/NCA fittings, supporting accurate and smooth operation in research infusion and perfusion setups.
Features
  • 1ft (30cm) length for flexible pump integration
  • .020in ID silicone tubing for low‑flow pumping
  • Straight‑through design for use with P720/NCA fittings
  • Non‑sterile tube sets supplied in packs of five
  • Compatible with all P720 peristaltic pump models
  • Ideal for precise infusion and perfusion applications