Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .031in ID silicone, straight through configuration (P720/NCA fittings required), 1ft (30cm) long. Package of 5, nonsterile.
SDP
Catalog No. 5081176
Encompass
The Instech P720/TS‑031S is a 1ft (30cm) straight‑through tube set for P720 peristaltic pumps, built with .031in ID silicone tubing to support mid‑range low‑flow applications. Used with P720/NCA fittings, these non‑sterile tube sets provide connector‑free inlet and outlet routing and are supplied in packs of five for reliable, smooth fluid delivery in research workflows.
Features
  • 1ft (30cm) length for flexible pump integration
  • .031in ID silicone tubing for mid‑range flow rates
  • Straight‑through design for use with P720/NCA fittings
  • Non‑sterile tube sets packaged in sets of five
  • Compatible with all P720 pump models
  • Supports precise infusion and perfusion applications
Catalog No. 50-811-76 Supplier Instech Supplier No. P720/TS031S
The Instech P720/TS‑031S is a 1ft (30cm) straight‑through tube set for P720 peristaltic pumps, built with .031in ID silicone tubing to support mid‑range low‑flow applications. Used with P720/NCA fittings, these non‑sterile tube sets provide connector‑free inlet and outlet routing and are supplied in packs of five for reliable, smooth fluid delivery in research workflows.
Features
  • 1ft (30cm) length for flexible pump integration
  • .031in ID silicone tubing for mid‑range flow rates
  • Straight‑through design for use with P720/NCA fittings
  • Non‑sterile tube sets packaged in sets of five
  • Compatible with all P720 pump models
  • Supports precise infusion and perfusion applications