Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .062in ID silicone, straight through configuration (P720/NCA fittings required), 1ft (30cm) long. Package of 5, nonsterile.
SDP
Catalog No. 5081177
Encompass
The Instech P720/TS‑093S is a 1ft (30cm) straight‑through tube set for P720 peristaltic pumps, made with .093in ID silicone tubing to support higher flow applications. Used with P720/NCA fittings, these non‑sterile tube sets provide connector‑free inlet and outlet routing and are supplied in packs of five for smooth and reliable fluid transfer in research workflows.
Features
  • 1ft (30cm) length for flexible pump integration
  • .093in ID silicone tubing for higher flow ranges
  • Straight‑through design for P720/NCA fittings
  • Packaged in sets of five, non‑sterile
  • Compatible with all P720 peristaltic pump models
Catalog No. 50-811-77 Supplier Instech Supplier No. P720/TS062S
The Instech P720/TS‑093S is a 1ft (30cm) straight‑through tube set for P720 peristaltic pumps, made with .093in ID silicone tubing to support higher flow applications. Used with P720/NCA fittings, these non‑sterile tube sets provide connector‑free inlet and outlet routing and are supplied in packs of five for smooth and reliable fluid transfer in research workflows.
Features
  • 1ft (30cm) length for flexible pump integration
  • .093in ID silicone tubing for higher flow ranges
  • Straight‑through design for P720/NCA fittings
  • Packaged in sets of five, non‑sterile
  • Compatible with all P720 peristaltic pump models