Learn More
Instech Tube sets for Instech P720 peristaltic pumps, .093in ID silicone, straight through configuration (P720/NCA fittings required), 1ft (30cm) long. Package of 5, nonsterile.
SDP
Catalog No. 5081178
Encompass
The Instech P720/TS‑093S is a 1ft (30cm) straight‑through tube set for P720 peristaltic pumps, using .093in ID silicone tubing to support the highest flow range in the P720 series. Designed for use with P720/NCA fittings, these non‑sterile tube sets provide connector‑free inlet and outlet routing and are supplied in packs of five for smooth, reliable fluid transfer in research applications.
Features
  • 1ft (30cm) straight‑through silicone tube set
  • .093in ID tubing for high‑flow P720 operation
  • Requires P720/NCA straight‑through fittings
  • Packaged in sets of five, non‑sterile
  • Compatible with all P720 peristaltic pump models
Catalog No. 50-811-78 Supplier Instech Supplier No. P720/TS093S
The Instech P720/TS‑093S is a 1ft (30cm) straight‑through tube set for P720 peristaltic pumps, using .093in ID silicone tubing to support the highest flow range in the P720 series. Designed for use with P720/NCA fittings, these non‑sterile tube sets provide connector‑free inlet and outlet routing and are supplied in packs of five for smooth, reliable fluid transfer in research applications.
Features
  • 1ft (30cm) straight‑through silicone tube set
  • .093in ID tubing for high‑flow P720 operation
  • Requires P720/NCA straight‑through fittings
  • Packaged in sets of five, non‑sterile
  • Compatible with all P720 peristaltic pump models