Learn More
Waters Corp Tube Shaker, 5 mL Adaptor, Vibration-Free, OneLab Connected
Catalog No. 502390905 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product integrates market-leading orbital shaker technology with OneLab software, allowing for remote setup and monitoring. It provides a wide range of vibration-free mixing speeds, aiming to minimize error and ensure maximum repeatability in laboratory processes.
  • Seamless connection with OneLab for remote setup and monitoring.
  • Offers a wide range of vibration-free mixing speeds.
  • Designed to minimize errors.
  • Ensures maximum repeatability.
  • Compatible with Andrew+, Pipette+, and manual systems.
  • Product type: Connected device.
Catalog No. 50-239-0905 Supplier Waters Corp Supplier No. 176005078
Add to Cart
Add to Cart
Request a Quote
This product integrates market-leading orbital shaker technology with OneLab software, allowing for remote setup and monitoring. It provides a wide range of vibration-free mixing speeds, aiming to minimize error and ensure maximum repeatability in laboratory processes.
  • Seamless connection with OneLab for remote setup and monitoring.
  • Offers a wide range of vibration-free mixing speeds.
  • Designed to minimize errors.
  • Ensures maximum repeatability.
  • Compatible with Andrew+, Pipette+, and manual systems.
  • Product type: Connected device.