Learn More
Andwin Scientific TUBE SIL MICRCNT 1.5ML PK500
SDP
Catalog No. NC9492494
Encompass_Preferred
Constructed from siliconized polypropylene, the 1.5 mL G-Tube Snap Cap Siliconized Microcentrifuge Tubes feature low surface adhesion and maximum sample recovery. The tubes are sold with caps that perform as secure seal for the tubes and also as a textured writing surface. This product is sold in a pack of 500.
Catalog No. NC9492494 Supplier Andwin Scientific Supplier No. 22179004
Constructed from siliconized polypropylene, the 1.5 mL G-Tube Snap Cap Siliconized Microcentrifuge Tubes feature low surface adhesion and maximum sample recovery. The tubes are sold with caps that perform as secure seal for the tubes and also as a textured writing surface. This product is sold in a pack of 500.