Learn More
Waters Corp Waters Corporation Tubing, Stainless Steel, 1/16 in O.D., 0.009 in I.D., 10 ft
Catalog No. 50818724 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This stainless steel tubing is designed for optimal performance in laboratory settings, ensuring leak-free couplings, minimal band spreading, and low dead volume. It provides essential flexibility in system setup and maintenance, contributing to enhanced productivity in scientific applications.
  • Designed for leak-free couplings, minimal band spreading, and low dead volume.
  • Features consistent diameter and precise ferrule geometry.
  • Offers flexibility in materials, lengths, and diameters.
  • Tested and certified to maintain peak system efficiency.
  • Guarantees consistent instrument operation and accurate results.
  • Manufactured in an ISO-certified facility using ultra-pure chemicals.
  • Each batch undergoes multiple tests for repeatable performance.
Catalog No. 50-818-724 Supplier Waters Corp Supplier No. WAT026973
This stainless steel tubing is designed for optimal performance in laboratory settings, ensuring leak-free couplings, minimal band spreading, and low dead volume. It provides essential flexibility in system setup and maintenance, contributing to enhanced productivity in scientific applications.
  • Designed for leak-free couplings, minimal band spreading, and low dead volume.
  • Features consistent diameter and precise ferrule geometry.
  • Offers flexibility in materials, lengths, and diameters.
  • Tested and certified to maintain peak system efficiency.
  • Guarantees consistent instrument operation and accurate results.
  • Manufactured in an ISO-certified facility using ultra-pure chemicals.
  • Each batch undergoes multiple tests for repeatable performance.