Learn More
Andwin Scientific TUBE SSPIN AST 1.5ML PK500
SDP
Catalog No. NC9891735
Encompass_Preferred
The 1.5 mL SuperSpin™ Microcentrifuge Tubes are ideal for vortexing and shaking tables. This product is able to endure centrifugations spins up to 35,000 g, which is twice the g-force most other microcentrifuge tubes can tolerate. The tubes are also
Catalog No. NC9891735 Supplier Andwin Scientific Supplier No. 20170022
The 1.5 mL SuperSpin™ Microcentrifuge Tubes are ideal for vortexing and shaking tables. This product is able to endure centrifugations spins up to 35,000 g, which is twice the g-force most other microcentrifuge tubes can tolerate. The tubes are also