Learn More
Electron Microscopy Sciences Stacking Platform for Tubes w/Dimpled Mat
SDP
Catalog No. 501929982
Encompass_Preferred
A stacking platform for use with the BenchBlotter 2D Fixed Speed Platform Rocker. 12 x 12", with dimpled mat. 3" separation.
Catalog No. 50-192-9982 Supplier Electron Microscopy Sciences Supplier No. 6570005
A stacking platform for use with the BenchBlotter 2D Fixed Speed Platform Rocker. 12 x 12", with dimpled mat. 3" separation.