Learn More
Ace Glass, Inc. Tube, storage, 210mL, 0-4mm Hi-Vac valve top with 3/8in cajon side port, No-Air
SDP
Supplier:  Ace Glass, Inc. 781315
Encompass_Preferred
210mL No-Air storage tube with 0-4mm Hi-vac valve top and a 9mm side port designed to accept 3/8 Cajon fittings. The valve at the top gives the researcher precise flow control.
Catalog No. 50-975-448