Learn More
Chemglass Life Sciences 350mL Tube, Storage, 14/20 Outer Joint, Airfree, Schlenk
SDP
Catalog No. 501530967
Encompass_Preferred
350mL Tube, Storage, 14/20 Outer Joint, Airfree, Schlenk. Similar to AF-0522, but cylindrical in shape. 0-8mm valve size. PLEASE NOTE: STIR BARS LARGER IN DIAMETER THAN THE OPENING OF THE VALVE, OR STANDARD TAPER JOINT, CAN BE SEALED INTO VESSELS DURING MANUFACTURING. PLEASE CALL FOR DETAILS.
Catalog No. 50-153-0967 Supplier Chemglass Life Sciences Supplier No. AF052304
350mL Tube, Storage, 14/20 Outer Joint, Airfree, Schlenk. Similar to AF-0522, but cylindrical in shape. 0-8mm valve size. PLEASE NOTE: STIR BARS LARGER IN DIAMETER THAN THE OPENING OF THE VALVE, OR STANDARD TAPER JOINT, CAN BE SEALED INTO VESSELS DURING MANUFACTURING. PLEASE CALL FOR DETAILS.