Learn More
Chemglass Life Sciences 50mL Tube, Storage, 24/40 Outer Joint, Airfree, Schlenk
SDP
Catalog No. 50873469
Encompass_Preferred
50mL Tube, Storage, 24/40 Outer Joint, Airfree, Schlenk. Similar to AF-0522, but cylindrical in shape. The 25 and 50mL sizes have a 0-4mm Chem-Vac™ Chem-Cap valve while the 100 and 350mL sizes have a 0-8mm valve size.
Catalog No. 50-873-469 Supplier Chemglass Life Sciences Supplier No. AF052306
50mL Tube, Storage, 24/40 Outer Joint, Airfree, Schlenk. Similar to AF-0522, but cylindrical in shape. The 25 and 50mL sizes have a 0-4mm Chem-Vac™ Chem-Cap valve while the 100 and 350mL sizes have a 0-8mm valve size.