Learn More
Boca Scientific Inc Tube Storage Box for 15ml Conical Tubes, Holds 36 Tubes, Set of 2
Catalog No. NC1671718
Encompass
These durable polypropylene tube storage boxes keep your 15ml samples safely stored and nicely organized. Each box has a 6 x 6 configuration and a capacity of 36 15ml conical tubes. Alpha-numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. These boxes are ideal for freezer and refrigerator storage, with a temperature rating of -90°C - 121°C. The boxes measure 5 5/8” x 5 5/8” x 5”h. They are sold in sets of two.
Catalog No. NC1671718 Supplier Boca Scientific Inc Supplier No. 27341
These durable polypropylene tube storage boxes keep your 15ml samples safely stored and nicely organized. Each box has a 6 x 6 configuration and a capacity of 36 15ml conical tubes. Alpha-numeric markings on the boxes provide a unique ID for every sample. The transparent cover allows you to see the samples and markings without removing the lid. These boxes are ideal for freezer and refrigerator storage, with a temperature rating of -90°C - 121°C. The boxes measure 5 5/8” x 5 5/8” x 5”h. They are sold in sets of two.