Learn More
Research Products International Corp Tube Storage Racks with Lid, 5 ml
SDP
Catalog No. 501978515
Encompass
Tube Storage Racks accept common micro-tube sizes including 0.5 ml, 1.5-2.0 ml, or 5.0 ml. Racks feature small footprint for bench and freezer storage and are ideal for holding screw cap tubes. Natural color racks are supplied with lids and feature side notches to facilitate easy lid removal. Withstand low temperatures down to -90°C and autoclaving. Supplied 5 racks per case.
  • Accomodate 0.5, 1.5-2.0 and 5.0 ml tubes
  • Withstand -90°C and autoclaving
Catalog No. 50-197-8515 Supplier Research Products International Corp Supplier No. 400220
Tube Storage Racks accept common micro-tube sizes including 0.5 ml, 1.5-2.0 ml, or 5.0 ml. Racks feature small footprint for bench and freezer storage and are ideal for holding screw cap tubes. Natural color racks are supplied with lids and feature side notches to facilitate easy lid removal. Withstand low temperatures down to -90°C and autoclaving. Supplied 5 racks per case.
  • Accomodate 0.5, 1.5-2.0 and 5.0 ml tubes
  • Withstand -90°C and autoclaving