Learn More
Bio Rad Laboratories TUBE STRIPS AND DOMED CAP STRI
Catalog No. 509010026
Encompass
Strip, Tube and Domed Cap; Bio-Rad; Natural; PCR; With caps; 20 bags of 12 8-tube strips and 12 8-cap strips; PCR volumes of 5-125uL recommended; 0.2mL
Catalog No. 50-901-0026 Supplier Bio Rad Laboratories Supplier No. TBC0802
Strip, Tube and Domed Cap; Bio-Rad; Natural; PCR; With caps; 20 bags of 12 8-tube strips and 12 8-cap strips; PCR volumes of 5-125uL recommended; 0.2mL