Learn More
Med Vet International HemaStop Styptic Tube, J1257, 3 Count
Catalog No. 5000495616
Encompass_Preferred
Veterinary styptics became necessary when the first nail trim started bleeding. Use onbleeding nails, beaks, skin, or any external minor bleeding. Hemastop is a combination of two proven styptic compounds:• Ferric subsulfate powder – industry standard• Petrolatum gel – used in the sport of boxing as a safe and pain free styptic for years.Both compounds are combined in a familiar ‘Chapstick’ type screw end dispenser with a protective cap.Economy – one tube has a large multitude of applications Ingredients – soy wax, petrolatum, ferric sulfate, diatomite Patent pendingHemastop. 3 tubes per pack
Catalog No. 50-004-95616 Supplier Med Vet International Supplier No. JOR1257
Veterinary styptics became necessary when the first nail trim started bleeding. Use onbleeding nails, beaks, skin, or any external minor bleeding. Hemastop is a combination of two proven styptic compounds:• Ferric subsulfate powder – industry standard• Petrolatum gel – used in the sport of boxing as a safe and pain free styptic for years.Both compounds are combined in a familiar ‘Chapstick’ type screw end dispenser with a protective cap.Economy – one tube has a large multitude of applications Ingredients – soy wax, petrolatum, ferric sulfate, diatomite Patent pendingHemastop. 3 tubes per pack