Learn More
Med Vet International McKesson Polypropylene Test Tube, 5 mL, 12 x 75 mm, 177-110441, 1000/Bag
Catalog No. 503044436
Encompass_Preferred
McKesson Test Tube Round Bottom Plain 12 X 75 mm 5 mL Without Color Coding Without Closure Polypropylene Tube 5 mL polystyrene translucent test tubes 12 x 75 millimeters Molded from premium-grade polystyrene (PS) for clarity Can withstand 1500 RPM during centrifugation Tubes are nongraduated, with round bottoms Brittleness temperature: 20° C Maximum temperature: 90° C 1,000 tubes per bag
Catalog No. 50-304-4436 Supplier Med Vet International Supplier No. 1082089BG
McKesson Test Tube Round Bottom Plain 12 X 75 mm 5 mL Without Color Coding Without Closure Polypropylene Tube 5 mL polystyrene translucent test tubes 12 x 75 millimeters Molded from premium-grade polystyrene (PS) for clarity Can withstand 1500 RPM during centrifugation Tubes are nongraduated, with round bottoms Brittleness temperature: 20° C Maximum temperature: 90° C 1,000 tubes per bag