Learn More
Perkin Elmer US LLC Quartz Sample Tube for high-temperature furnace
Catalog No. 509044733
Encompass_Preferred
Required for the expansion, penetration, compression and flexure modes of operation. Used with high temperature furnace, part of Quartz Parallel Plate System Kit (Product Number: N5390138)
Catalog No. 50-904-4733 Supplier Perkin Elmer US LLC Supplier No. N5191549
Required for the expansion, penetration, compression and flexure modes of operation. Used with high temperature furnace, part of Quartz Parallel Plate System Kit (Product Number: N5390138)